Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3320

Sigma-Aldrich

IL-36RA human

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)

Synonym(s):

FIL1 delta, IL-1F5, IL-1HY1, IL-1L1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$357.00

$357.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 μG
$357.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$357.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

17.0 kDa

packaging

pkg of 25 μg

impurities

endotoxin, tested

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL36RN(26525)

General description

The IL-1 family is comprised of 11 structurally related ligands, including the recently re-named IL-36RA (IL-1F5), IL-36? (IL-1F6), IL-36? (IL-1F8), and IL-36? (IL-1F9). The interaction of IL-36 ligands with the IL-1Rrp2 receptor (IL-1R6) can induce various activities, including dendritic cell maturation and activation. IL-36RA can antagonize the NF-kappaB signaling induced by either IL-36?, -? or -? by binding to the IL-1Rrp2 receptor in a manner that prevents the initiation of functional signaling. Recombinant human IL-36RA is an E. coli derived 17 kDa protein containing 154 amino acid residues.

Biochem/physiol Actions

The IL-1 family is comprised of 11 structurally related ligands, including the recently re-named IL-36RA (IL-1F5), IL-36? (IL-1F6), IL-36? (IL-1F8), and IL-36? (IL-1F9). Recombinant human IL-36RA is an E. coli derived 17 kDa protein containing 154 amino acid residues.

Physical form

Lyophilized from 10 mM Sodium Phosphate, pH 7.5.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jennifer E Towne et al.
The Journal of biological chemistry, 286(49), 42594-42602 (2011-10-04)
IL-36α, IL-36β, and IL-36γ (formerly IL-1F6, IL-1F8, and IL-1F9) are IL-1 family members that signal through the IL-1 receptor family members IL-1Rrp2 (IL-1RL2) and IL-1RAcP. IL-36Ra (formerly IL-1F5) has been reported to antagonize IL-36γ. However, our previous attempts to demonstrate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service