Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP3074

Sigma-Aldrich

IL-12p40 human

recombinant, expressed in CHO cells, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in CHO cells

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

40 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL12B(3593)

General description

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Accumulating data indicate that p40 secretion precedes that of IL-12 expression. In addition, to its ability to covalently bind to p35 to form IL-12, p40 can bind to p19 to form IL-23. Recombinant human IL-12 p40 is a 40.0 kDa protein containing 306 amino acid residues.
The interleukin-12 subunit β (IL12β) gene is localized on human chromosome 5q31–33. The encoded protein is part of IL-12, which is a heterodimer. It is mainly expressed by antigen-presenting cells (APCs).

Biochem/physiol Actions

IL-12 is a disulfide-linked heterodimeric protein (p70), composed of two subunits, p35 and p40, which are encoded by two different genes. Recombinant human IL-12 p40 is a 40.0 kDa protein containing 306 amino acid residues.
Interleukin-12 subunit β (IL12β) is an immunoregulatory cytokine. It is involved in pathogenic defense responses and is also suggested as an anticancer therapeutic agent. IL12B is responsible for polarizing naive CD4+ cells toward a TH1 (T helper) phenotype and inducing IFN-γ (interferon) secretion by T and natural killer (NK) cells. This protein also stimulates the differentiation of cytotoxic CD8+ cells and enhances the activity of NK cells. IL12B is linked with many autoimmune diseases, such as psoriasis, systemic lupus erythematosus, Crohn′s disease and ulcerative colitis.

Sequence

p40 Subunit: IWELKKDVYV VELDWYPDAP GEMVVLTCDT PEEDGITWTL DQSSEVLGSG KTLTIQVKEF GDAGQYTCHK GGEVLSHSLL LLHKKEDGIW STDILKDQKE PKNKTFLRCE AKNYSGRFTC WWLTTISTDL TFSVKSSRGS SDPQGVTCGA ATLSAERVRG DNKEYEYSVE CQEDSACPAA EESLPIEVMV DAVHKLKYEN YTSSFFIRDI IKPDPPKNLQ LKPLKNSRQV EVSWEYPDTW STPHSYFSLT FCVQVQGKSK REKKDRVFTD KTSATVICRK NASISVRAQD RYYSSSWSEW ASVPCS

Physical form

Lyophilized from 1x PBS, pH 7.2.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in 1x PBS, pH 7.2 to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Interleukin-12 Levels May Prevent an Increase in the Amount of Fungi in the Gastrointestinal Tract during the First Years of Diabetes Mellitus Type 1.
Kowalewska B, et al.
Disease Markers (2016)
Interleukin-12 Immunomodulation Delays the Onset of Lethal Peritoneal Disease of Ovarian Cancer.
Cohen CA, et al.
Journal of Interferon & Cytokine Research, 36(1), 62-73 (2016)
Li Zhang et al.
PloS one, 10(6), e0130982-e0130982 (2015-06-24)
This study aims to determine whether the genetic polymorphisms of IL-12B gene is a susceptibility factor to Ankylosing spondylitis (AS) in mainland Han Chinese population. Eight single-nucleotide polymorphisms (SNPs) (rs10045431, rs11167764, rs3212227, rs6556412, rs6556416, rs6871626, rs6887695 and rs7709212) in the
M K Gately et al.
Journal of immunology (Baltimore, Md. : 1950), 147(3), 874-882 (1991-08-01)
IL-12 is a heterodimeric cytokine that was identified on the basis of its ability to synergize with IL-2 in the induction of cytotoxic effector cells and was originally called cytotoxic lymphocyte maturation factor (CLMF). IL-12 was also found to stimulate
M B Atkins et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 3(3), 409-417 (1997-03-01)
A Phase I dose escalation trial of i.v. administered recombinant human interleukin 12 (rhIL-12) was performed to determine its toxicity, maximum tolerated dose (MTD), pharmacokinetics, and biological and potential antineoplastic effects. Cohorts of four to six patients with advanced cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service