Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2132

Sigma-Aldrich

RAR, γ isoform, GST tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE)

Synonym(s):

NR1B3, RARC

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$422.00

$422.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
$422.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$422.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

frozen liquid

mol wt

~77 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

800 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RARG(5916)

Biochem/physiol Actions

Nuclear receptors form the largest known family of transcription factors and have a crucial role in nearly all aspects of vertebrate development and adult physiology by transducing the effects of hormones into transcriptional responses. The family is defined by two domains: (a) the central, highly conserved, DNA-binding domain (DBD) of approx. 66 amino acids, and (b) the C-terminal, structurally conserved, ligand-binding domain (LBD) of approx. 250 amino acids. The amino-terminal regions are least conserved among nuclear receptor sequences. Retinoids, the natural and syntheic derivatives of vitamin A, have been shown to inhibit the growth of many human tumor cells. Although the exact mechanism for this growth suppression remains unknown, the importance of retinoic acid receptors (RARs) and retinoid X receptors (RXRs) have been established in a number of tumor cell models. Three types of RARs have been identified (RAR, RAR, and RAR ) with homologous but slightly different retinoic acid (RA) binding domains. Transactivation assays and retinoid binding assays with RAR, RAR, and RAR have shown that each RAR has a different affinity for RA and RA analogs.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retinoic acid receptors: transcription factors modulating gene regulation, development, and differentiation.
E Linney
Current topics in developmental biology, 27, 309-350 (1992-01-01)
M Leid et al.
Trends in biochemical sciences, 17(10), 427-433 (1992-10-01)
Complexity in the retinoid signalling system arises from a combination of several forms of retinoic acid, multiple cytoplasmic binding proteins and nuclear receptors, and the existence of polymorphic retinoic acid response elements. Additional diversity appears to be generated by heterodimeric
Nonsteroid nuclear receptors: what are genetic studies telling us about their role in real life?
P Kastner et al.
Cell, 83(6), 859-869 (1995-12-15)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service