Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP2067

Sigma-Aldrich

CAR, ligand binding domain (101-348) human

recombinant, expressed in E. coli, ≥85% (SDS-PAGE)

Synonym(s):

CAR, CAR1, MB67, MGC150433, MGC97144, MGC97209

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

biological source

human

recombinant

expressed in E. coli

Assay

≥85% (SDS-PAGE)

form

frozen liquid

mol wt

~30 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

500 μg/mL

color

clear
colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... NR1I3(9970)

Biochem/physiol Actions

The constitutive androstane receptor (CAR) was identified as a member of the orphan nuclear hormone receptor family in 1994. Although constitutively active, it can be further activated by `phenobarbitol-like` compounds, the most potent being the synthetic compound TCPOBOP. Upon activation, CAR regulates the xenobiotic drug metabolizing enzymes, cytochrome P450s. There are several overlapping functions between the nuclear receptors PXR and CAR. Recently, CAR, as well as PXR, have been shown to place a role in bile acid clearance and cholestatic liver injury.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Baes et al.
Molecular and cellular biology, 14(3), 1544-1552 (1994-03-01)
We have identified and characterized a new orphan member of the nuclear hormone receptor superfamily, called MB67, which is predominantly expressed in liver. MB67 binds and transactivates the retinoic acid response elements that control expression of the retinoic acid receptor
P Wei et al.
Nature, 407(6806), 920-923 (2000-11-01)
Organisms encounter a wide range of foreign compounds--or 'xenobiotics'--with potentially harmful consequences. The cytochrome P450 (CYP) enzymes metabolize xenobiotics and thus are a primary defence against these compounds. Increased expression of specific CYP genes in response to particular xenobiotics is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service