Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP2050

Sigma-Aldrich

RAR, β human

recombinant, expressed in E. coli, ≥75% (SDS-PAGE)

Synonym(s):

HAP, NR1B2, RRB2

Sign Into View Organizational & Contract Pricing

Select a Size

5 μG
$604.00

$604.00


Please contact Customer Service for Availability


Select a Size

Change View
5 μG
$604.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$604.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in E. coli

Assay

≥75% (SDS-PAGE)

form

frozen liquid

mol wt

~52.5 kDa

packaging

pkg of 5 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

250 μg/mL

color

clear colorless

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... RARB(5915)

Biochem/physiol Actions

Retinoic acid receptors are important in the regulation of growth and differentiation of epithelial tissues, embryonic and central nervous system development and hematopoiesis. Retinoids mediate their effect by two classes of nuclear receptor proteins, the retinoic acid receptors (RARs) and the retinoid X receptors (RXRs), that each consist of three isotypes ( α, β, and γ) encoded in separate genes. Upon dimerization with RXR, RARs can bind to specific enhancer sequences in the DNA, so-called retinoic acid response elements (RAREs), resulting in transcriptional activation of target genes in the presence of ligand. Retinoids, the natural and synthetic vitamin A derivatives, are known to inhibit the proliferation of lung cancer and breast cancer cells and the growth of carcinogen-induced bronchogenic squamous cell carcinoma and mammary tumors, and have been used as chemoprevention agents against both types of cancer. A growing body of evidence supports the hypotheses that the RAR beta gene is a tumor suppressor gene and the chemopreventive effects of retinoids are due to induction of RAR beta. RAR beta expression is reduced in many malignant tumors including breast carcinoma.

Physical form

Clear and colorless frozen liquid solution

Preparation Note

Use a manual defrost freezer and avoid repeated freeze-thaw cycles. While working, please keep sample on ice.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Retinoic acid receptors: transcription factors modulating gene regulation, development, and differentiation.
E Linney
Current topics in developmental biology, 27, 309-350 (1992-01-01)
Nonsteroid nuclear receptors: what are genetic studies telling us about their role in real life?
P Kastner et al.
Cell, 83(6), 859-869 (1995-12-15)
M Leid et al.
Trends in biochemical sciences, 17(10), 427-433 (1992-10-01)
Complexity in the retinoid signalling system arises from a combination of several forms of retinoic acid, multiple cytoplasmic binding proteins and nuclear receptors, and the existence of polymorphic retinoic acid response elements. Additional diversity appears to be generated by heterodimeric

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service