Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0664

Sigma-Aldrich

CBL-B, GST-Tag

recombinant, expressed in E. coli

Synonym(s):

Casitas B-lineage lymphoma proto-oncogene b (CBL-B), E3 ubiquitin-protein ligase CBL-B, RING finger protein 56

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$624.00

$624.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μG
$624.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$624.00


Please contact Customer Service for Availability

recombinant

expressed in E. coli

form

frozen liquid

mol wt

72 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CBLB(868)

General description

Human Casitas B-lineage lymphoma proto-oncogene b (CBL-B), also known as E3 ubiquitin-protein ligase CBL-B, RING finger protein 56, and SH3-binding protein CBL-B with Y363F mutation GenBank Accession No. NM_017934, a.a. 39 - 426 with N-terminal GST-Tag expressed in an E. coli expression system. MW=72 kDa.

Physical form

Aqueous solution

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service