Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0609

Sigma-Aldrich

TUT4, FLAG® -Tag, Biotin Labeled

recombinant, expressed in HEK 293 cells

Synonym(s):

TUT4, Terminal uridylyltransferase 4, ZCCHC11

Sign Into View Organizational & Contract Pricing

Select a Size

25 μG
$657.00

$657.00


Please contact Customer Service for Availability


Select a Size

Change View
25 μG
$657.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

$657.00


Please contact Customer Service for Availability

recombinant

expressed in HEK 293 cells

form

frozen liquid

mol wt

127 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ZCCHC11(23318)

General description

Human TUT4, also known as ZCCHC11, GenBank Accession number NM_001009881, a.a. 212-1313 with C-terminal FLAG-Avi-tag. MW = 127 kDa, expressed in an HEK293 expression system. This protein is enzymatically biotinylated using Avitag technology. Biotinylation is confirmed to be ?90%.

Physical form

Aqueous solution

Legal Information

AviTag is a trademark of Avidity, LLC
FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service