Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP0524

Sigma-Aldrich

COX2, FLAG® -His-Tags

recombinant, expressed in baculovirus infected Sf9 cells

Synonym(s):

Cyclooxygenase-2 Prostaglandin-Endo-peroxide Synthase 2, HCox-2, Inducible Prostaglandin H Synthase 2, PHS-2, PTGS2, Prostaglandin G/H Synthase and Cyclooxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.26

recombinant

expressed in baculovirus infected Sf9 cells

form

frozen liquid

mol wt

71 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... COX2(6775079)

General description

Human COX2, GenBank Accession No. NM_000963, a.a. 1-604 (end), with C-terminal FLAG-His-tag, MW=71 kDa, expressed in a baculovirus infected Sf9 cell expression system.

Physical form

Aqueous solution

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service