Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0483

Sigma-Aldrich

CREBBP (1081-1197) His tag human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

CBP, CREB binding protein, KAT3A, RSTS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

14.9 kDa

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

1.26 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... CREBBP(1387)

General description

Human bromodomain CREB binding protein (CREBBP), also known as CBP, RSTS and KAT3A (GenBank Accession No. NM_004380) amino acids 1081- 1197 with N-terminal His-tag, MW=14.9 kDa, expressed in an E. coli expression system.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei-Chien Huang et al.
Molecular cell, 26(1), 75-87 (2007-04-17)
CBP plays a central role in coordinating and integrating multiple signaling pathways. Competition between NF-kappaB and p53 for CBP is a crucial determinant of whether a cell proliferates or undergoes apoptosis. However, how the CBP-dependent crosstalk between these two transcription
I Panagopoulos et al.
Human molecular genetics, 10(4), 395-404 (2001-02-07)
The CBP gene at 16p13 fuses to MOZ and MLL as a result of the t(8;16)(p11;p13) in acute (myelo)monocytic leukemias (AML M4/M5) and the t(11;16)(q23;p13) in treatment-related AML, respectively. We show here that a novel t(10;16)(q22;p13) in a childhood AML

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service