Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0474

Sigma-Aldrich

ATAD2A (981-1108) human

recombinant, expressed in E. coli, ≥90% (SDS-PAGE)

Synonym(s):

ANCCA, ATPase family, AAA domain containing 2, PRO2000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥90% (SDS-PAGE)

form

aqueous solution

mol wt

16.2 kDa

packaging

pkg of 100 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ATAD2(29028)

General description

Human Bromodomain ATAD2A (GenBank Accession No. NM_014109) amino acids 981 – 1108 with N-terminal HIS-tag, MW = 16.2 kDa, expressed in an E. coli expression system.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ekaterina V Kalashnikova et al.
Cancer research, 70(22), 9402-9412 (2010-09-25)
Chromatin coregulators are important factors in tumorigenesis and cancer progression. ANCCA is an AAA+ ATPase and a bromodomain-containing nuclear coactivator for the estrogen and androgen receptors that is crucial for assembly of chromatin-modifying complexes and proliferation of hormone-responsive cancer cells.
C Caron et al.
Oncogene, 29(37), 5171-5181 (2010-06-29)
Cancer cells frequently express genes normally active in male germ cells. ATAD2 is one of them encoding a conserved factor harbouring an AAA type ATPase domain and a bromodomain. We show here that ATAD2 is highly expressed in testis as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service