Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0429

Sigma-Aldrich

PDE4A10 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥75% (SDS-PAGE)

Synonym(s):

DPDE2, PDE46, phosphodiesterase 4A, cAMP-specific

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected Sf9 cells

Assay

≥75% (SDS-PAGE)

form

aqueous solution

specific activity

≥75% (compared to standard)

mol wt

117 kDa

packaging

pkg of 10 μg

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE4A(5141)

General description

Human PDE4A10, also known as PDE4A protein isoform 6, transcript variant 3 (GenBank Accession No. NM_001111309) amino acids 2-825 (end) with N-terminal GST-tag, MW= 117 kDa, expressed in a Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole of 3?, 5?-cAMP to 5?-AMP per min at 37°C.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I McPhee et al.
Cellular signalling, 13(12), 911-918 (2001-12-01)
In situ hybridisation methods were used to map the distribution of the novel long PDE4A10 isoform in the brain. PDE4A10 distribution was compared to that of the long PDE4A5 isoform and the short PDE4A1 isoform using probes specific for unique
Angela McCahill et al.
Cellular signalling, 20(11), 2071-2083 (2008-08-30)
Transcripts for the PDE4A10 cyclic AMP phosphodiesterase isoform are present in a wide variety of rat tissues including the heart. Sequence comparisons between the putative human and mouse promoters revealed a number of conserved regions including both an Sp1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service