Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0384

Sigma-Aldrich

MLL1/WDR5/Ash2L/RbBP5/DPY30 human

recombinant, expressed in E. coli, ≥83% (SDS-PAGE)

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
$840.00

$840.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
$840.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$840.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

recombinant

expressed in E. coli

Assay

≥83% (SDS-PAGE)

form

aqueous solution

mol wt

12 kDa (DPY30)
27 kDa (MLL1)
35 kDa (WDR5)
60 kDa (RbBP5)
61 kDa (Ash2L)

packaging

pkg of 50 μg

shipped in

dry ice

storage temp.

−70°C

General description

MLL1/WARD complex of human MLL1 (GenBank Accession No. NM_005933) amino acids 3745-end with N-terminal His-tag, MW= 27 kDa, human WDR5 (GenBank Accession No. NM_017588), amino acids 22-end with N-terminal Flagtag, MW= 35 kDa, human Ash2L (GenBank Accession No. NM_001105214), amino acids 2-end with N-terminal His-tag, MW= 61 kDa, human RbBP5 (GenBank Accession No. NM_005057), amino acids 2-end with N-terminal His-tag, MW= 60 kDa, and human DPY30 (GenBank Accession No. NM_032574), amino acids 2-end with N-terminal His-tag, MW=12 kDa, individually expressed in an E. coli expression system.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Man Mohan et al.
Nature reviews. Cancer, 10(10), 721-728 (2010-09-17)
The RNA polymerase II (Pol II) elongation factor (ELL) was the first translocation partner of mixed lineage leukaemia (MLL) for which a biochemical function was determined. It was therefore proposed that the regulation of the elongation stage of transcription could
Anamika Patel et al.
The Journal of biological chemistry, 284(36), 24242-24256 (2009-06-27)
Transcription in eukaryotic genomes depends on enzymes that regulate the degree of histone H3 lysine 4 (H3K4) methylation. The mixed lineage leukemia protein-1 (MLL1) is a member of the SET1 family of H3K4 methyltransferases and is frequently rearranged in acute

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service