Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRP0257

Sigma-Aldrich

PDE1A Active human

recombinant, expressed in baculovirus infected insect cells, ≥70% (SDS-PAGE)

Synonym(s):

Calcium/calmodulin-dependent 3′,5′-cyclic nucleotide phosphodiesterase 1A, HDAM1, HSPDE1A, Phosphodiesterase 1A

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥70% (SDS-PAGE)

form

aqueous solution

specific activity

≥1237 units/μg

mol wt

88.3 kDa

packaging

pkg of 10 μg

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PDE1A(5136)

General description

Human PDE1A (GenBank Accession No. NM_005019), full length with N-terminal GST tag, MW = 88.3 kDa, expressed in a Baculovirus infected Sf9 cell expression system

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Unit Definition

One unit is defined as the amount of enzyme that will convert 1 pmole of 3′, 5′-cAMP to 5′ AMP per min at 37°C

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 20% glycerol and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service