Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SRP0243

Sigma-Aldrich

aRAF Active human

recombinant, expressed in baculovirus infected insect cells, ≥80% (SDS-PAGE)

Synonym(s):

A-Raf proto-oncogene, PKS2, v-Raf murine sarcoma 3611 viral oncogene homolog

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
$516.00

$516.00


Please contact Customer Service for Availability


Select a Size

Change View
10 μG
$516.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

$516.00


Please contact Customer Service for Availability

biological source

human

recombinant

expressed in baculovirus infected insect cells

Assay

≥80% (SDS-PAGE)

form

aqueous solution

mol wt

63.6 kDa

packaging

pkg of 10 μg

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... ARAF(369)

General description

Human A-RAF (GenBank Accession No. NM_001654), amino acids 282-end, containing the mutations YY301-302DD, MW = 63.6 kDa, with N-terminal GST tag, expressed in an Baculovirus infected Sf9 cell expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Physical form

Formulated in 25 mM Tris-HCl, pH 8.0, 100 mM NaCl, 0.05% Tween-20, 50% glycerol, and 3 mM DTT.

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube containing enzyme to recover full content of the tube. Aliquot enzyme into single use aliquots. Store remaining undiluted enzyme in aliquots at -70°C. Note: Enzyme is very sensitive to freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service