Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRP0160

Sigma-Aldrich

Histone H3 peptide (21-44) human

≥90% (HPLC)

Synonym(s):

HIST1H3E, Histone H3.1

Sign Into View Organizational & Contract Pricing

Select a Size

160 NMOL
$252.00

$252.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
160 NMOL
$252.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

$252.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

human

Assay

≥90% (HPLC)

form

aqueous solution

mol wt

~2 kDa

packaging

pkg of 160 nmol

storage condition

avoid repeated freeze/thaw cycles

concentration

>0.02 mg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... HIST1H3E(8353)

General description

A peptide substrate for Histone 3 K27 methyltransferases.

Application

Study enzyme kinetics, and screen small molecular inhibitors of Histone 3 K27 methyltransferases for drug discovery and HTS applications

Physical form

Supplied as water solution, 800 μM

Preparation Note

Thaw on ice. Upon first thaw, briefly spin tube to recover full content of the tube. Aliquot into single use aliquots. Store remaining product in aliquots at -70°C. Note: Avoid freeze/thaw cycles.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service