Skip to Content
MilliporeSigma
All Photos(1)

Documents

SRE0086

Sigma-Aldrich

Lysis Solution for Blood

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105600

form

liquid

Quality Level

storage temp.

room temp

General description

The Lysis Solution for Blood is used together with the Neutralization Solution for Blood (SRE0087) to rapidly extract and neutralize DNA from whole blood, whole blood dried on a blood card, and cultured mammalian cells.

Other Notes

Stable for two years from the date of manufacture when stored at room temperature. Do not use past expiration date printed on product label.

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service