Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRE0076

Sigma-Aldrich

SeqClin Wash Solution, 5X Concentrate

Reagent designed and manufactured under cGMP controls suitable for use in an IVD application

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352201
NACRES:
NA.25

Quality Level

form

liquid

application(s)

diagnostic assay manufacturing

storage temp.

room temp

Application

SeqClin Wash Solution is recommended for use in a maintenance wash of sequencing systems.

Physical form

This 5X SeqClin stock solution contains 2.5% TWEEN® 20 with 0.15% ProClin 300. Preparation of 1X SeqClin working buffer: Dilute the 5X concentrated buffer 5-fold with ultrapure water (= 18 MΩ x cm resistivity at 25 °C).

Notes: If precipitation is present in the 5X concentrated buffer, warm the bottle to 25 °C and mix until
completely dissolved prior to dilution. It is recommended 1X working solutions be filtered through a 0.2 mm filter before use.

Legal Information

ProClin is a trademark of LANXESS Corp.
SeqClin is a trademark of Sigma-Aldrich Co. LLC
TWEEN is a registered trademark of Croda International PLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - Skin Sens. 1

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service