Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SRE0029

Sigma-Aldrich

Cardiolipin solution from bovine heart

4.7-5.3 mg/mL in ethanol, ≥97% (TLC), Suitable for manufacturing of diagnostic kits and reagents

Synonym(s):

Cardiolipin solution from bovine heart

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$337.00
500 MG
$2,070.00
1000 MG
$3,680.00

$337.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
$337.00
500 MG
$2,070.00
1000 MG
$3,680.00

About This Item

EC Number:
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

$337.00


Please contact Customer Service for Availability

Request a Bulk Order

biological source

bovine heart

Quality Level

Assay

≥97% (TLC)

form

liquid

concentration

4.7-5.3 mg/mL in ethanol

lipid type

phosphoglycerides

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application


  • Site of action of the local anesthetic tetracaine in a phosphatidylcholine bilayer with incorporated cardiolipin.: This paper examines how tetracaine interacts with phosphatidylcholine bilayers containing cardiolipin, which has implications for understanding local anesthetic mechanisms (Shibata et al., 1995).


Biochem/physiol Actions

Cardiolipin is a mitochondrial phospholipid that is found in mammalian tissues in low concentrations. It is often a membrane component and can inhibit cell attachment of fibronectin, vitronectin, and type I collagen.

Physical form

Approximately 5 mg/ml in ethanol.

Other Notes

>80% polyunsaturated fatty acid content, primarily linoleic acid.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup - Solvent

Flash Point(C)

9.7 °C - closed cup - Solvent


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service