Skip to Content
MilliporeSigma
All Photos(5)

Documents

SRE0020

Sigma-Aldrich

Butyrylcholinesterase from equine serum

Suitable for manufacturing of diagnostic kits and reagents, ≥900 units/mg protein

Synonym(s):

Butyrylcholinesterase from equine serum, Acylcholine acyl-hydrolase, Choline esterase, butyryl, Pseudocholinesterase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204

Quality Level

form

lyophilized powder

specific activity

≥900 units/mg protein

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Unit Definition

One unit will hydrolyze 1.0 micromole of butyrylcholine to choline and butyrate per minute at pH 8.0 at 37 deg C.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service