Skip to Content
MilliporeSigma
All Photos(2)

Documents

SRE0018

Sigma-Aldrich

Lyticase from Arthrobacter Luteus

Suitable for manufacturing of diagnostic kits and reagents, lyophilized powder, ≥2000 units/mg protein

Synonym(s):

Lyticase from Arthrobacter luteus

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.32

biological source

bacterial (Arthrobacter luteus)

Quality Level

form

lyophilized powder

specific activity

≥2000 units/mg protein

composition

Protein, ≥20% biuret

application(s)

diagnostic assay manufacturing

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Lyticase hydrolyzes poly-β(1→3)-glucose such as yeast cell wall glucan.

Unit Definition

One unit will produce a ΔA800 of 0.001 per min at pH 7.5 at 25 °C, using a suspension of yeast as substrate in a 3 mL reaction mixture.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Louis Dacquay et al.
G3 (Bethesda, Md.), 7(6), 1799-1809 (2017-04-30)
Actively proliferating cells constantly monitor and readjust their metabolic pathways to ensure the replenishment of phospholipids necessary for membrane biogenesis and intracellular trafficking. In Saccharomyces cerevisiae, multiple studies have suggested that the lysine acetyltransferase complex NuA4 plays a role in

Protocols

This procedure may be used for the determination of Lyticase activity using Baker’s yeast as the substrate.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service