Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4072

Tetrahydroberberine

new

≥98% (HPLC)

Synonym(s):

(±)-Canadine, (±)-Tetrahydroberberine, 5,8,13,13a-tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-alpha]quinolizine, Canadine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21NO4
CAS Number:
Molecular Weight:
339.39
MDL number:
UNSPSC Code:
12352200

biological source

(Corydalis yanhusuo)

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

N21C(Cc5c(c(c(cc5)OC)OC)C2)c3c(cc4c(c3)OCO4)CC1

InChI key

VZTUIEROBZXUFA-UHFFFAOYSA-N

Biochem/physiol Actions

Dopamine receptor antagonist; nicotinic acetylcholine receptor (α4β2, α4β4, α7, α1β, 1γδ) modulator; induces clearance of low-density lipoprotein in mice.

Tetrahydroberberine (Canadine), an isoquinoline alkaloid isolated from corydalis tuber, is a potent dopamine D2 receptor antagonist (pKi = 6.08) and nicotinic acetylcholine receptor (α4β2, α4β4, α7, α1β, 1γδ) modulator. Tetrahydroberberine alleviates high-fat diet-induced hyperlipidemia and induces clearance of low-density lipoprotein and intermediate-density lipoprotein in mice.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guilan Wei et al.
European journal of pharmacology, 968, 176433-176433 (2024-02-19)
The promotion of excess low-density lipoprotein (LDL) clearance stands as an effective clinical approach for treating hyperlipidemia. Tetrahydroberberine, a metabolite of berberine, exhibits superior bioavailability compared to berberine and demonstrates a pronounced hypolipidemic effect. Despite these characteristics, the impact of
Tae Ho Lee et al.
The Journal of pharmacology and experimental therapeutics, 338(3), 917-924 (2011-06-11)
Because delayed gastric emptying and impaired gastric accommodation are regarded as pathophysiological mechanisms underlying functional dyspepsia (FD), prokinetics and fundic relaxants have been suggested as a new treatment for FD. We isolated tetrahydroberberine (THB), an isoquinoline alkaloid (5,8,13,13a-tetrahydro-9,10-dimethoxy-6H-benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine) from Corydalis
Erika Plazas et al.
Pharmacological research, 177, 106126-106126 (2022-02-14)
Complex diseases such as neurodegenerative disorders and cancer constitute a growing public health problem due to the rising incidence and lack in effective therapies. Since pharmacotherapy based on a single target has been insufficient for drug development in complex diseases

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service