Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4031

Sigma-Aldrich

BAX Dimer Modulator 19, BDM19

new

≥98% (HPLC)

Synonym(s):

BDM-19, (E)-4-(6-Iodo-2-(3-methylstyryl)-4-oxoquinazolin-3(4H)-yl)benzoic acid, 4-[6-Iodo-2-[2-(3-methylphenyl)ethenyl]-4-oxo-3(4H)-quinazolinyl]benzoic acid, BDM 19, BDM19

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H17IN2O3
CAS Number:
Molecular Weight:
508.31
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

IC1=CC=C2C(C(N(C(/C=C/C3=CC(C)=CC=C3)=N2)C4=CC=C(C=C4)C(O)=O)=O)=C1

InChI key

CQEADRMBOXGOAE-LFYBBSHMSA-N

Biochem/physiol Actions

Potent and selective cytosolic BAX dimer modulator that effectively induces apoptosis in cancer cells by selectively modulating inactive BAX.

BDM19, a potent BAX dimer modulator with a Kd of 560 nM, selectively binds to the N-terminal trigger site of inactive BAX, thereby inhibiting its dimerization with an IC50 of 5 μM. By stabilizing the inactive conformation of BAX, BDM-19 allosterically activates it, targeting cells containing cytosolic inactive BAX monomers and dimers. This leads to a dose-dependent translocation of BAX to mitochondria, subsequent cytochrome c release, and induction of apoptosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nadege Gitego et al.
Nature communications, 14(1), 8381-8381 (2023-12-17)
The BCL-2 family protein BAX is a major regulator of physiological and pathological cell death. BAX predominantly resides in the cytosol in a quiescent state and upon stress, it undergoes conformational activation and mitochondrial translocation leading to mitochondrial outer membrane

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service