Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML4025

Sigma-Aldrich

Drp1 inhibitor, SC9

new

≥98% (HPLC)

Synonym(s):

N-[6-Amino-1,2,3,4-tetrahydro-2,4-dioxo-1-(phenylmethyl)-5-pyrimidinyl]-2-[(5,7-dimethyl[1,2,4]triazolo[1,5-a]pyrimidin-2-yl)thio]-N-methylacetamide, SC9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N8O3S
CAS Number:
Molecular Weight:
466.52
MDL number:
UNSPSC Code:
12352200
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

CC1=NC2=NC(SCC(N(C(C(N3)=O)=C(N)N(CC4=CC=CC=C4)C3=O)C)=O)=NN2C(C)=C1

Biochem/physiol Actions

Cell penetrant, potent and selective inhibitor of GTPase activity of dynamin-related protein 1 (Drp1).
Drp1 inhibitor SC9 is a cell penetrant, potent and selective inhibitor of GTPase activity of dynamin-related protein 1 (Drp1) that binds to a switch I-adjacent grove (SWAG) on Drp1. SC9 inhibits Drp1-Fis1 interaction in cells and eases the burden of Drp1-mediated pathologies such as LPS-induced mitochondrial dysfunction. It rescues mice from LPS-induced endotoxemia.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis Rios et al.
Nature communications, 14(1), 4356-4356 (2023-07-20)
The large cytosolic GTPase, dynamin-related protein 1 (Drp1), mediates both physiological and pathological mitochondrial fission. Cell stress triggers Drp1 binding to mitochondrial Fis1 and subsequently, mitochondrial fragmentation, ROS production, metabolic collapse, and cell death. Because Drp1 also mediates physiological fission

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service