Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3977

proTAME

new

≥90% (HPLC)

Synonym(s):

(2S)-2-[[(4-Methylphenyl)sulfonyl]amino]-9,13-dioxo-14-phenyl-7-[[[[(2-phenylacetyl)oxy]methoxy]carbonyl]amino]-10,12-dioxa-6,8-diazatetradec-6-enoic acid methyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

SML3977-1MG
$275.00
SML3977-5MG
$1,110.00

$275.00


Please contact Customer Service for Availability


Select a Size

Change View
SML3977-1MG
$275.00
SML3977-5MG
$1,110.00

About This Item

Empirical Formula (Hill Notation):
C34H38N4O12S
CAS Number:
Molecular Weight:
726.75
UNSPSC Code:
12352200

$275.00


Please contact Customer Service for Availability

Quality Level

Assay

≥90% (HPLC)

form

(Powder or solid or crytals or semi-solid)

storage condition

desiccated

color

colorless to yellow

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(CC1=CC=CC=C1)OCOC(/N=C(NC(OCOC(CC2=CC=CC=C2)=O)=O)\NCCC[C@H](NS(C3=CC=C(C=C3)C)(=O)=O)C(OC)=O)=O

Biochem/physiol Actions

Cell-permeable N,N′-bis(acyloxymethyl carbamate) prodrug of the anaphase-promoting complex (APC) inhibitor Nα-p-Tosyl-L-arginine methyl ester (TAME).
proTAME is a cell-permeable N,N′-bis(acyloxymethyl carbamate) prodrug of the anaphase-promoting complex (APC) inhibitor Nα-p-Tosyl-L-arginine methyl ester (TAME). Upon entering cells, proTAME (12 μM) is effectively converted by cellular esterases to TAME, which in turn inactivates the spindle assembly checkpoint (SAC) by blocking APC-dependent proteolysis, causing prolonged mitotic duration at low doses (780 nM - 3 μM in HeLa cells, 6 μM in hTERT-RPE1 cells) or mitotic arrest and cell death at high doses (12 μM in HeLa cells). Note: proTAME is not active in some cells (e.g. MCF10A, due to the lack of esterase activity required for proTAME activation.

Caution

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ProTAME Arrest in Mammalian Oocytes and Embryos Does Not Require Spindle Assembly Checkpoint Activity.
International Journal of Molecular Sciences, 20(18), 4537-4537 (2019)
Xing Zeng et al.
Cancer cell, 18(4), 382-395 (2010-10-19)
Microtubule inhibitors are important cancer drugs that induce mitotic arrest by activating the spindle assembly checkpoint (SAC), which, in turn, inhibits the ubiquitin ligase activity of the anaphase-promoting complex (APC). Here, we report a small molecule, tosyl-L-arginine methyl ester (TAME)
Pauline Douglas et al.
Molecular and cellular biology, 40(13) (2020-04-15)
The DNA-dependent protein kinase catalytic subunit (DNA-PKcs) has well-established roles in DNA double-strand break repair, and recently, nonrepair functions have also been reported. To better understand its cellular functions, we deleted DNA-PKcs from HeLa and A549 cells using CRISPR/Cas9. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service