Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3963

Sigma-Aldrich

DPPA-1 acetate

≥95% (HPLC)

Synonym(s):

(D)-APP acetate, (D)-NYSKPTDRQYHF acetate, (D)-PPA 1 acetate, (D)-PPA acetate, (D)-PPA-1 acetate, (D)-PPA1 acetate, Asn-Tyr-Ser-Lys-Pro-Thr-Asp-Arg-Gln-Tyr-His-Phe (all D-amino acids) acetate, D PD-1/PD-L1 interaction Antagonist-1 acetate, D-APP acetate, D-Asn-D-Tyr-D-Ser-D-Lys-D-Pro-D-Thr-D-Asp-D-Arg-D-Gln-D-Tyr-D-His-D-Phe, D-PPA 1 acetate, D-PPA acetate, D-type APP acetate, D-type anti-PD-L1 peptide acetate, DPPA 1 acetate, DPPA-1 acetate, DPPA1 acetate, NYSKPTDRQYHF (all D-amino acids) acetate, PD-L1-targeting D-peptide acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C70H98N20O21 · xC2H4O2
Molecular Weight:
1555.65 (free base basis)
UNSPSC Code:
51111800
UNSPSC Code:
12352200

Quality Level

Assay

≥95% (HPLC)

form

(Powder or Lyophilized powder or film)

storage condition

desiccated

color

white to off-white

storage temp.

-10 to -25°C

Biochem/physiol Actions

Programmed death-1 (PD-1) and programmed death-ligand 1 (PD-L1) interaction antagonist/blocker with anti-cancer efficacy in vitro and in vivo.



DPPA-1 is a programmed cell death protein 1 (PD-1) and PD-1 ligand (PD-L1) interaction antagonist/blocker that targets PD-L1 via direct affinity binding (KD = 0.51 μM). DPPA-1 effectively blocks soluble Fc-labeled human PD-L1 from binding cell surface PD-1 in vitro and shows anti-tumor efficacy among CT26 tumor-bearing mice in vivo (2 mg/kg via daily i.p.).

Citation

Hygroscopic

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service