Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3946

Sigma-Aldrich

DFPQ

≥98% (HPLC)

Synonym(s):

AP-06-202, N4-Cyclohexyl-N2-(3,4-difluorophenyl)-2,4-quinazolinediamine, N4-Cyclohexyl-N2-(3,4-difluorophenyl)quinazoline-2,4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H20F2N4
CAS Number:
Molecular Weight:
354.40
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Gs-biased, β2 subtype-selective adrenergic receptor (AR) negative allosteric modulator (NAM) that inhibits β-arrestin recruitment, but not Gs coupling to β2AR.

DFPQ is a Gs-biased, β2 subtype-selective adrenergic receptor (AR) negative allosteric modulator (NAM) that inhibits β-arrestin recruitment (IC50 = 600 nM; 30-min DFPQ pretreatment prior to 12-min 1 μM ISO induction), but not Gs coupling to β2AR (30-min 10 μM DFPQ pretreatment prior to 10-min 1 μM ISO-induced cAMP production). DFPQ effectively inhibits agonist-induced β2AR phosphorylation (IC50 = 2.6 μM; 30-min DFPQ pretreatment prior to 10-min 1 μM ISO induction) and internalization (100% inhibition by 30-min 10 μM DFPQ prior to 1h 10 μM ISO) and protects against the functional desensitization of β-agonist-mediated regulation in cell and tissue models with minimal effects on β1AR.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Ippolito et al.
Proceedings of the National Academy of Sciences of the United States of America, 120(31), e2302668120-e2302668120 (2023-07-25)
Catecholamine-stimulated β2-adrenergic receptor (β2AR) signaling via the canonical Gs-adenylyl cyclase-cAMP-PKA pathway regulates numerous physiological functions, including the therapeutic effects of exogenous β-agonists in the treatment of airway disease. β2AR signaling is tightly regulated by GRKs and β-arrestins, which together promote

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service