Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3867

Sigma-Aldrich

BI, Broccoli Fluorophore

≥98% (HPLC)

Synonym(s):

(5Z)-3-(1H-Benzimidazol-7-ylmethyl)-5-[(3,5-difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2-methyl-4H-imidazol-4-one, (Z)-3-((1H-Benzo[d]imadazol-4-yl)methyl)-5-(3,5-difluoro-4-hydroxybenzylidene)-2-methyl-3,5-dihydro-4H-imidazol-4-one, BI, DFHBI-BI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14F2N4O2
CAS Number:
Molecular Weight:
368.34
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

Biochem/physiol Actions

Cell-permeable DFHBI derivative that selectively binds to Broccoli aptamer; used for imaging of RNA in living cells.



BI is a cell-permeable DFHBI benzimidazolo (DFHBI-BI) derivative that greatly and selectively promotes and stabilizes Broccoli aptamer folding in vitro and cells with enhanced photostability. BI displays little background in vitro and cells (ϕ=0.00048, ϵ=42,500 M−1 cm−1), but very high fluorescence activation after binding to Broccoli (ϕ=0.669, ϵ=33,600 M−1 cm−1, KD = 51 nM). DFHBI-BI shows negligible cellular toxicity and binds Squash with much weaker affinity than Broccoli. Ex/Em: 470/505 nm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fluorophore-Promoted RNA Folding and Photostability Enables Imaging of Single Broccoli-Tagged mRNAs in Live Mammalian Cells
Angewandte Chemie (International ed. in English), 59(11), 4511-4518 (2020)
Single mRNA Imaging with Fluorogenic RNA Aptamers and Small-molecule Fluorophores
Wei Chen, Xiaoying Zhao, Nanyang Yang, Xing Li
Angewandte Chemie (International Edition in English), 62(7) (2023)
Sourav Kumar Dey et al.
Nature chemical biology, 18(2), 180-190 (2021-12-24)
Fluorogenic RNA aptamers are used to genetically encode fluorescent RNA and to construct RNA-based metabolite sensors. Unlike naturally occurring aptamers that efficiently fold and undergo metabolite-induced conformational changes, fluorogenic aptamers can exhibit poor folding, which limits their cellular fluorescence. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service