Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3826

Sigma-Aldrich

LW-3-254

≥98% (HPLC)

Synonym(s):

(3aR,4R,4aR,5aS,6S,6aS)-2-(4-(2-(3,5-dichlorophenyl)-2-oxoethoxy)phenyl)-4,4a,5,5a,6,6a-hexahydro-4,6-ethenocyclopropa[f]isoindole-1,3(2H,3aH)-dione, Analog 13

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19Cl2NO4
Molecular Weight:
468.33
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

LW-3-254, a derivative of MM0299, is an orally bioavailable and blood-brain barrier penetrant, selective and potent inhibitor of lanosterol synthase (LSS) that exhibits anti-glioblastoma activity. LW-3-254 stimulates 24(S),25-epoxycholesterol (EPC) synthesis in GBM tumors, which induces toxicity in glioma stem-like cells. It potently inhibits Mut6 (Glioblastoma stem-like cells) cell growth by depleting cellular cholesterol.
Orally bioavailable and blood-brain barrier penetrant, selective and potent inhibitor of lanosterol synthase (LSS)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective and brain-penetrant lanosterol synthase inhibitors target glioma stem-like cells by inducing 24(S),25-epoxycholesterol production
Cell Chemical Biology, 30(2), 214-229 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service