Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3813

Sigma-Aldrich

YKL-05-099

new

≥98% (HPLC)

Synonym(s):

3-(2-Chloro-6-methylphenyl)-7-((2-methoxy-4-(1-methylpiperidin-4-yl)phenyl)amino)-1-(5-methoxypyridin-2-yl)-3,4-dihydropyrimido[4,5-d]pyrimidin-2(1H)-one, YKL-05-99

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H34ClN7O3
CAS Number:
Molecular Weight:
600.11
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Faint yellow to yellow-brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=CC=CC(Cl)=C1N2C(N(C3=CC=C(OC)C=N3)C4=NC(NC5=C(OC)C=C(C6CCN(C)CC6)C=C5)=NC=C4C2)=O

Biochem/physiol Actions

Potent and selective salt-inducible kinases (SIK1, SIK2, SIK3) inhibitor in vitro and in vivo.



YKL-05-099 is a potent and selective salt-inducible kinases inhibitor (SIK1/2/3 IC50 = 10/40/30 nM) with superior pharmacokinetic properties than HG-9-91-01. YKL-05-099 pre-treatment of murine BDMCs up-regulates Zymosan A-stimulated IL-10 (EC50 = 460 nM with 24h ), while suppresses inflammatory cytokines production (TNFα, IL-6 and IL-12p40). Likewise, YKL-05-099 intraperitoneal administration in mice (20 mg/kg i.p.) 15 min prior to LPS (0.5 mg/kg i.p.) enhances serum IL-10, while reduces HDAC5 pSer259 levels in total splenic leukocytes and serum TNFα level in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service