Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3777

Sigma-Aldrich

SAR7334

≥98% (HPLC)

Synonym(s):

SAR-7334, 4-({(1R,2R)-2-[(3R)-3-Aminopiperidin-1-yl]-2,3-dihydro-1H-inden-1-yl}oxy)-3-chlorobenzonitrile, 4-[rac-trans-(1,2)-2-((R)-3-Aminopiperidin-1-yl)indan-1-yloxy]-3-chlorobenzonitrile, SAR 7334

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22ClN3O
CAS Number:
Molecular Weight:
367.87
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

Clc1c(ccc(c1)C#N)O[C@H]2[C@@H](Cc4c2cccc4)N3C[C@@H](CCC3)N

InChI key

RLKRLNQEXBPQGQ-OZOXKJRCSA-N

Biochem/physiol Actions

TRPC6-selective transient receptor potential channel (TRPC) inhibitor.



SAR7334 is a TRPC6-selective transient receptor potential channel (TRPC) inhibitor (IC50 against OAG-induced Ca2+ influx in TRPC3/6/7-expressing cells = 9.5/226/282 nM, IC50 >10 μM in TRPC4/10 cells; IC50 = 7.9 nM by whole-cell patch-clamp using TRPC6-HEK-FITR cells).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and pharmacological characterization of a novel potent inhibitor of diacylglycerol-sensitive TRPC cation channels
British Journal of Pharmacology, 172(14), 3650-3660 (2015)
The role of TRPC6 calcium channels and P2 purinergic receptors in podocyte mechanical and metabolic sensing
Physiology international (2021)
TRPC6 inhibits renal tubular epithelial cell pyroptosis through regulating zinc influx and alleviates renal ischemia-reperfusion injury
Faseb Journal, 36(10) (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service