Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3766

Sigma-Aldrich

SRI-42127

≥98% (HPLC)

Synonym(s):

3-(1H-Indazol-6-yl)-N-methyl-N-(tetrahydro-2H-pyran-4-yl)imidazo[1,2-b]pyridazin-6-amine

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$245.00
25 MG
$986.00

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$245.00
25 MG
$986.00

About This Item

Empirical Formula (Hill Notation):
C19H20N6O
CAS Number:
Molecular Weight:
348.40
UNSPSC Code:
12352200
NACRES:
NA.77

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

Biochem/physiol Actions

Blood-brain barrier permeable HuR cytoplasmic dimerization inhibitor that reduces the stability of targeted mRNAs
SRI-42127 is a blood-brain barrier permeable imidazopyridazinamine analog that potently and reversibly inhibits the RNA-binding protein HuR multimerization and blocks HuR nucleocytoplasmic shuttling. SRI-42127 suppresses the expression and release of several pro-inflammatory mediators with minimal effect on anti-inflammatory cytokines. SRI 42127 weakens HuR interaction with Bcl-2 and Mcl-1 leading to induction of apoptosis and decreased tumor growth. SRI42127 subdues microglial activation in vivo and attenuates the recruitment/chemotaxis of neutrophils and monocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of the RNA Regulator HuR by SRI-42127 Attenuates Neuropathic Pain After Nerve Injury Through Suppression of Neuroinflammatory Responses
Neurotherapeutics, 19(5), 1649-1661 (2022)
Targeting the HuR Oncogenic Role with a New Class of Cytoplasmic Dimerization Inhibitors
Cancer research, 81(8), 2220-2233 (2021)
SRI-42127, a novel small molecule inhibitor of the RNA regulator HuR, potently attenuates glial activation in a model of lipopolysaccharide-induced neuroinflammation
Glia, 70(1), 155-172 (2022)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service