Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3693

Sigma-Aldrich

RA839

≥98% (HPLC)

Synonym(s):

(3S)-1-[4-[[(2,3,5,6-Tetramethylphenyl)sulfonyl]amino]-1-naphthalenyl]-3-pyrrolidinecarboxylic acid, RA 839

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N2O4S
CAS Number:
Molecular Weight:
452.57
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

RA839 is an active in vivo, highly selective and potent activator of Nrf2 (transcription factor NF-E2-related factor 2) signaling. RA839 binds noncovalently to the Nrf2-interacting kelch domain of Keap1 thereby inhibiting the interaction between Keap1 and Nrf2. RA839 inhibits the induction of both inducible nitric oxide synthase expression and nitric oxide release in response to lipopolysaccharides in macrophages. RA-839 is a potent antagonist of Rotavirus propagation in vitro that inhibits viral RNA and protein expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Upayan Patra et al.
Antiviral research, 161, 53-62 (2018-11-23)
Acute watery diarrhea due to Rotavirus (RV) infection is associated with high infantile morbidity and mortality in countries with compromised socio-economic backgrounds. Although showing promising trends in developed countries, the efficacy of currently licensed RV vaccines is sub-optimal in socio-economically
Kim T Tran et al.
Journal of medicinal chemistry, 62(17), 8028-8052 (2019-08-15)
Inhibiting the protein-protein interaction (PPI) between the transcription factor Nrf2 and its repressor protein Keap1 has emerged as a promising strategy to target oxidative stress in diseases, including central nervous system (CNS) disorders. Numerous non-covalent small-molecule Keap1-Nrf2 PPI inhibitors have
Angelika F Winkel et al.
The Journal of biological chemistry, 290(47), 28446-28455 (2015-10-16)
The activation of the transcription factor NF-E2-related factor 2 (Nrf2) maintains cellular homeostasis in response to oxidative stress by the regulation of multiple cytoprotective genes. Without stressors, the activity of Nrf2 is inhibited by its interaction with the Keap1 (kelch-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service