Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SML3625

Sigma-Aldrich

BAY-069

≥98% (HPLC)

Synonym(s):

3-(4-Chloro-3-(o-tolyloxy)naphthalen-1-yl)-6-(trifluoromethyl)pyrimidine-2,4(1H,3H)-dione, 3-(5-chloro-4-(2-methyl-phenoxy)-bicyclo[4.4.0]deca-1(6),2,4,7,9-pentaen-2-yl)-6-(trifluoro-methyl)-1,2,3,4-tetrahydro-pyrimidine-2,4-dione, BAY 069, BAY069

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$245.00
25 MG
$986.00

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$245.00
25 MG
$986.00

About This Item

Empirical Formula (Hill Notation):
C22H14ClF3N2O3
CAS Number:
Molecular Weight:
446.81
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$245.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

shipped in

wet ice

storage temp.

−70°C

Biochem/physiol Actions

BAY-069 is a non-cytotoxic (anti-proliferation IC50 >47 μM using 6 cell lines), potent and selective branched chain amino acid (BCAA) transaminase (BCAT) inhibitor (BCAT1/2 IC50 = 27/130 nM; glutamic-oxaloacetic transaminase GOT1/2 IC50 >50 μM) with good selectivity over a panel of 30 kinases, 30 proteases and 77 other targets. BAY-069 upregulates BCAAs in high BCAT1-expressing U-87 MG and high BCAT2-expressing MDA-MB-231 (IC50 = 358 and 874 nM, respectively) with good pharmacokinetics and oral bioavailability in rats in vivo.
Orally available, non-cytotoxic, potent and selective branched chain amino acid (BCAA) transaminase 1/2 (BCAT1 & BCAT2) inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service