Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3583

Sigma-Aldrich

PF-04634817 succinate

≥98% (HPLC)

Synonym(s):

1,​5-​Anhydro-​2,​3-​dideoxy-​4-​O-​methyl-​3-​[[(1R,​3S)​-​3-​(1-​methylethyl)​-​3-​[[(1S,​4S)​-​5-​[6-​(trifluoromethyl)​-​4-​pyrimidinyl]​-​2,​5-​diazabicyclo[2.2.1]​hept-​2-​yl]​carbonyl]​cyclopentyl]​amino]​-D-​erythro-​pentitol succinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H36F3N5O3 · C4H6O4
CAS Number:
Molecular Weight:
629.67
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

room temp

SMILES string

CO[C@H]1[C@@H](N[C@@H]2CC[C@@](C(N3C[C@@]4([H])N(C5=NC=NC(C(F)(F)F)=C5)C[C@]3([H])C4)=O)(C(C)C)C2)CCOC1

InChI key

MCRWZBYTLVCCJJ-DKALBXGISA-N

Biochem/physiol Actions

Orally available, potent and selective CCR2/5 receptor antagonist

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of PF-04634817, an Oral CCR2/5 Chemokine Receptor Antagonist, on Albuminuria in Adults with Overt Diabetic Nephropathy
Gale JD, Gilbert S, Blumenthal S, Elliott T, Pergola PE, Goteti K, Scheele W, Perros-Huguet C
Kidney international reports, 3, 1316-1327 (2018)
Identification of Molecular Signatures and Candidate Drugs in Vascular Dementia by Bioinformatics Analyses
Shu J, Wei W, Zhang L
Frontiers in Molecular Neuroscience, 15, 751044-751044 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service