Skip to Content
MilliporeSigma
All Photos(2)

Documents

SML3558

Sigma-Aldrich

CQ211

≥98% (HPLC)

Synonym(s):

1,5-Dihydro-8-(6-methoxy-3-pyridinyl)-1-[4-(1-piperazinyl)-3-(trifluoromethyl)phenyl]- 4H -1,2,3-triazolo[4,5-c ]quinolin-4-one, 8‑(6-Methoxypyridin-3-yl)-1-(4-(piperazin-1-yl)-3-(trifluoromethyl)phenyl)-1,5-dihydro-4H-[1,2,3]triazolo[4,5‑c]quinolin-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22F3N7O2
CAS Number:
Molecular Weight:
521.49
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1C2=C(C3=CC(C4=CC=C(N=C4)OC)=CC=C3N1)N(N=N2)C5=CC=C(C(C(F)(F)F)=C5)N6CCNCC6

Biochem/physiol Actions

CQ211 is a cell penetrant, selective and highly potent RIOK2 (Right open reading frame kinase 2) inhibitor. CQ211 potently inhibits proliferation of multiple cancer cell lines including MKN-1 and HT-29 cells. It inhibits tumor progression in MKN-1 xenograft model after intraperitoneal administration.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yifan Ouyang et al.
Journal of medicinal chemistry, 65(11), 7833-7842 (2022-05-19)
RIOK2 is an atypical kinase implicated in multiple human cancers. Although recent studies establish the role of RIOK2 in ribosome maturation and cell cycle progression, its biological functions remain poorly elucidated, hindering the potential to explore RIOK2 as a therapeutic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service