Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3521

Sigma-Aldrich

m-Trifluoromethyl-diphenyl diselenide

≥98% (HPLC)

Synonym(s):

1,2-Bis(3-(trifluoromethyl)phenyl)diselane, Bis(3-trifluoromethylphenyl) diselenide, Di(3-trifluoromethylphenyl) diselenide, TFDD, [(m-CF3-PhSe)2; Bis[3-(trifluoromethyl)phenyl] diselenide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$112.00
25 MG
$452.00

$112.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$112.00
25 MG
$452.00

About This Item

Empirical Formula (Hill Notation):
C14H8F6Se2
CAS Number:
Molecular Weight:
448.12
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.25

$112.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

oil

color

colorless to yellow

storage temp.

-10 to -25°C

SMILES string

FC(F)(C1=CC([Se][Se]C2=CC=CC(C(F)(F)F)=C2)=CC=C1)F

InChI key

DGOYKERNPXVRDM-UHFFFAOYSA-N

Biochem/physiol Actions

Brain blood barrier penetrant organoselenium compound that exhibits antinociceptive and antidepressant effects
m-Trifluoromethyl-diphenyl diselenide (TFDD) is a brain blood barrier penetrant organoselenium compound that exhibits antinociceptive and antidepressant effects in animal models without development of tolerance or withdrawal sighs. m-Trifluoromethyl-diphenyl diselenide acts as opioid system modulator, which attenuates morphine withdrawal signs in mice. It exhibits antioxidant and anti-inflammatory properties which might contribute to neuro protective effects.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cleisson Schossler Garcia et al.
Molecular neurobiology, 58(10), 5078-5089 (2021-07-11)
Chronic pain and depression often coexist sharing common pathological mechanisms, and available analgesics and antidepressants have demonstrated limited clinical efficacy. Evidence has demonstrated that neuronal oxidative stress, apoptosis, and also glucocorticoid receptor dysregulation facilitate the occurrence and development of both
Contribution of Opioid and Nitrergic Systems to m-Trifluoromethyl diphenyl Diselenide Attenuates Morphine-Induced Tolerance in Mice
ACS Chemical Neuroscience, 13(7), 910-919 (2022)
Carolina C Martins et al.
Progress in neuro-psychopharmacology & biological psychiatry, 98, 109803-109803 (2019-11-07)
The opioid withdrawal syndrome is defined as a complex phenomenon involving multiple cellular adaptations, which leads to the emergence of aversive physical and affective signs. The m-trifluoromethyl-diphenyl diselenide (m-CF3-PhSe)2 elicits an antidepressant-like effect by modulating the opioid system in different

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service