Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3496

Sigma-Aldrich

MI-463

≥98% (HPLC)

Synonym(s):

4-Methyl-5-[[4-[[6-(2,2,2-trifluoroethyl)thieno[2,3-d]pyrimidin-4-yl]amino]-1-piperidinyl]methyl]-1H-indole-2-carbonitrile, MI 463, MI463

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H23F3N6S
CAS Number:
Molecular Weight:
484.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N#CC1=CC2=C(C)C(CN3CCC(CC3)NC4=C5C=C(CC(F)(F)F)SC5=NC=N4)=CC=C2N1

Biochem/physiol Actions

MI-463 is an orally active and potent menin-MLL interaction inhibitor (cell-free IC50 = 15.3 nM against 1 nM FITC-MLL(4-13) peptide) that targets menin MLL-binding site with high affinity (KD = 9.9 nM by ITC). MI-463 selectively inhibits mixed lineage leukemia (MLL) oncogenic fusion-driven cancer progression in cultures (GI50 by MTT = 230 nM vs. >5 μM against murine BMCs with or without MLL-AF9 fusion transformation) and is efficacious against human MLL leukemia MV4;11 tumor growth in mice in vivo (35 mg/kg/day i.p.) and improve the survival of MLL leukemic mice (40-50 mg/kg b.i.d. p.o.).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service