Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3457

Sigma-Aldrich

JX06

≥98% (HPLC)

Synonym(s):

4,4′-(Dithiodicarbonothioyl)bis[morpholine], 4-Morpholinethiocarbonyl disulfide, Bis(4-morpholinethiocarbonyl) disulfide, Bis(oxydi-2,1-ethanediyl)-thiuram disulfide, Dimorpholinethiuram disulfide, JX 06, JX-06, NSC 402538

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16N2O2S4
CAS Number:
Molecular Weight:
324.51
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

InChI

1S/C10H16N2O2S4/c15-9(11-1-5-13-6-2-11)17-18-10(16)12-3-7-14-8-4-12/h1-8H2

InChI key

KKVYOWPPMNSLCP-UHFFFAOYSA-N

Biochem/physiol Actions

JX06 is a potent, PDK1-selective, covalent pyruvate dehydrogenase (PDH) kinase (PDHK, PDK) inhibitor (PDK1/2/3 IC50 = 49/101/313 nM; PDK4 IC50 >10 µM, IC50 >10 µM against >300 other kinases). JX06 inhibits cellular PDK substrate PDHA1 Ser293/232 phosphorylation, resulting in reduced aerobic glycolysis and cancer proliferation in cultures (1-10 µM) and in mice in vivo (40-80 mg/kg via daily i.p. injection; A549 & HT-29 xenografts). JX06 is shown to target PDK1 Cys240 for covalent modification.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service