Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3451

Sigma-Aldrich

Maytansine

≥95% (HPLC)

Synonym(s):

MTS, Maitansine, Maysanine, Maytansin, N-Acetyl-N-methyl-L-alanine [1S-(1R*,2S*,3R*,5R*,6R*,16E,18E,20S*,21R*)]-11-chloro-21-hydroxy-12,20-dimethoxy-2,5,9,16-tetramethyl-8,23-dioxo-4,24-dioxa-9,22-diazatetracyclo[19.3.1.110,14.03,5]hexacosa-10,12,14(26),16,18-pentaen-6-yl ester, NSC 153858

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$120.00
25 MG
$485.00

$120.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$120.00
25 MG
$485.00

About This Item

Empirical Formula (Hill Notation):
C34H46ClN3O10
CAS Number:
Molecular Weight:
692.20
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$120.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O[C@]([C@@H](/C=C/C=C(CC1=CC2=C(C(OC)=C1)Cl)\C)OC)(C[C@H](O3)[C@H]([C@H]4[C@](C)([C@H](CC(N2C)=O)OC([C@@H](N(C)C(C)=O)C)=O)O4)C)NC3=O

InChI

1S/C34H46ClN3O10/c1-18-11-10-12-26(45-9)34(43)17-25(46-32(42)36-34)19(2)30-33(5,48-30)27(47-31(41)20(3)37(6)21(4)39)16-28(40)38(7)23-14-22(13-18)15-24(44-8)29(23)35/h10-12,14-15,19-20,25-27,30,43H,13,16-17H2,1-9H3,(H,36,42)/b12-10+,18-11+/t19-,20+,25+,26-,27+,30+,33+,34+/m1/s1

InChI key

WKPWGQKGSOKKOO-RSFHAFMBSA-N

Biochem/physiol Actions

Cytotoxic agent, microtubule inhibitor

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Muta. 2 - Repr. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengyang Jiang et al.
Bioconjugate chemistry, 29(9), 2920-2926 (2018-08-14)
This study was undertaken to target cell surface receptors other than the ones typically associated with breast cancer {estrogen receptor (ER), progesterone receptor (PR), and human epidermal growth factor receptor 2 (HER2)}. It was also launched to use small molecules
Grégory Menchon et al.
Nature communications, 9(1), 2106-2106 (2018-05-31)
Microtubule-targeting agents (MTAs) like taxol and vinblastine are among the most successful chemotherapeutic drugs against cancer. Here, we describe a fluorescence anisotropy-based assay that specifically probes for ligands targeting the recently discovered maytansine site of tubulin. Using this assay, we have
Maxine Bauzon et al.
Oncoimmunology, 8(4), e1565859-e1565859 (2019-03-25)
Oncology treatment has been revolutionized by the introduction of immune checkpoint inhibitor drugs, which enable 20-40% of patients to generate anti-tumor immune responses. Combination treatment approaches with chemotherapeutic drugs may enable responses in the remaining patient cohorts. In this regard

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service