Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3432

Sigma-Aldrich

Brevianamide F

≥98% (HPLC)

Synonym(s):

(3S,8aS)-hexahydro-3-(1H-indol-3-ylmethyl)-pyrrolo[1,2-a]pyrazine-1,4-dione, cyclo-(L-Trp-L-Pro), cyclo-(tryptophanyl-prolyl)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$87.80
25 MG
$353.00

$87.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$87.80
25 MG
$353.00

About This Item

Empirical Formula (Hill Notation):
C16H17N3O2
CAS Number:
Molecular Weight:
283.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

$87.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

N21[C@@H](CCC2)C(=O)N[C@H](C1=O)Cc3c4c([nH]c3)cccc4

InChI

1S/C16H17N3O2/c20-15-14-6-3-7-19(14)16(21)13(18-15)8-10-9-17-12-5-2-1-4-11(10)12/h1-2,4-5,9,13-14,17H,3,6-8H2,(H,18,20)/t13-,14-/m0/s1

InChI key

RYFZBPVMVYTEKZ-KBPBESRZSA-N

Biochem/physiol Actions

Brevianamide F is a naturally occurring cyclic dipeptide diketopiperazine (DKP) with antibacterial activity. It has shown activity against M. luteus and against methicillin-sensitive and resistant S. aureus among others.
Naturally occurring alkaloid with antibacterial activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Muhanna M Alshaibani et al.
Drug design, development and therapy, 10, 1817-1827 (2016-06-23)
Zingiber spectabile, commonly known as Beehive Ginger, is used as an ethnobotanical plant in many countries as an appetizer or to treat stomachache, toothache, muscle sprain, and as a cure for swelling, sores and cuts. This is the first report
Pei Huang et al.
Applied microbiology and biotechnology, 100(17), 7437-7447 (2016-03-16)
Microbes belonging to the genus Verrucosispora possess significant chemical diversity and biological properties. They have attracted the interests of many researchers and are becoming promising resources in the marine natural product research field. A bioassay-guided isolation from the crude extract
Abigail L Lind et al.
mSphere, 3(2) (2018-03-23)
Biosynthesis of many ecologically important secondary metabolites (SMs) in filamentous fungi is controlled by several global transcriptional regulators, like the chromatin modifier LaeA, and tied to both development and vegetative growth. In Aspergillus molds, asexual development is regulated by the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service