Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3426

Sigma-Aldrich

Glycerol phenylbutyrate

≥95% (HPLC)

Synonym(s):

1,2,3-Propanetriyl tris(4-phenylbutanoate), GPB, Glyceryl tri-[4-phenylbutyrate, HPN 100, Tri(4-phenylbutyryl)glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C33H38O6
CAS Number:
Molecular Weight:
530.65
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

oil

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O=C(OCC(OC(CCCC1=CC=CC=C1)=O)COC(CCCC2=CC=CC=C2)=O)CCCC3=CC=CC=C3

Biochem/physiol Actions

Glycerol phenylbutyrate, a triacylglycerol, is a prodrug of ER stress inhibitor 4-phenylbutyric acid (PBA). Glycerol phenylbutyrate is a nitrogen-binding agent approved for the chronic management of adult and pediatric patients (≥2 years of age) with urea cycle disorders (UCDs) who cannot be managed by dietary protein restriction and/or amino acid supplementation alone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service