Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3403

Sigma-Aldrich

HSip-1 DA trifluoroacetate hydrate

≥95% (HPLC)

Synonym(s):

Cyclen-AF-Cu DA trifluoroacetate hydrate, HSip 1 DA, HSip-1 diacetate trifluoroacetate hydrate, HSip1 DA, Hydrogen Sulfide imaging probe-1 DA, [N-(3′,6′-bis(Acetyloxy)-3-oxospiro[isobenzofuran-1(3H),9′-[9H]xanthen]-5-yl)-1,4,7,10-tetraazacyclododecane-1-acetamide-κN1,κN4,κN7,κN10]-copper(2+) trifluoroacetate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H37CuN5O8· xCF3COOH· yH2O
UNSPSC Code:
41106609
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

powder

mol wt

707.23 g/mol

storage condition

protect from light

color

, Light Blue to Green Blue

solubility

H2O: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C1OC2(C3=CC=C(NC(C[N]45CCN6CCN7CCN([Cu@+2]765)CC4)=O)C=C13)C8=CC=C(C=C8OC9=CC(OC(C)=O)=CC=C92)OC(C)=O.[y H2O]

Biochem/physiol Actions

Cell-permeable HSip-1 diacetate precursor. Sensitive and selective intracellular hydrogen sulfide (H2S) fluorescent probe.
HSip-1 DA corresponds to the cell-permeable diacetate ester precursor of the sensitive hydrogen sulfide (H2S)-selective fluorescent turn-on probe HSip-1 (Cyclen-AF-Cu; Hydrogen Sulfide imaging probe-1). Once entering cells, HSip-1 DA is hydrolyzed by esterases to form HSip-1, allowing the selective detection of intracellular H2S by fluorescence (Ex 470-490 nm, Em 510-550 nm). For intracellular loading, incubate cells with 5-30 μM Hsip-1 DA in DMEM at 37 °C for 30 min before removing unincorporated probe by washing with HBSS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service