Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3362

Sigma-Aldrich

YM-244769 dihydrochloride

≥98% (HPLC)

Synonym(s):

N-​[(3-​Aminophenyl)​methyl]​-​6-​[4-​[(3-​fluorophenyl)​methoxy]​phenoxy]​-3-​pyridinecarboxamide dihydrochloride, N-(3-Aminobenzyl)-6-{4-[3-fluorobenzyl)oxy]phenoxy} nicotinamide dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H22FN3O3·2HCl
CAS Number:
Molecular Weight:
516.39
MDL number:
UNSPSC Code:
51111800
UNSPSC Code:
12352200
NACRES:
NA.21
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to pink

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

O=C(C1=CC=C(OC2=CC=C(OCC3=CC=CC(F)=C3)C=C2)N=C1)NCC4=CC=CC(N)=C4

InChI

1S/C26H22FN3O3/c27-21-5-1-4-19(13-21)17-32-23-8-10-24(11-9-23)33-25-12-7-20(16-29-25)26(31)30-15-18-3-2-6-22(28)14-18/h1-14,16H,15,17,28H2,(H,30,31)

InChI key

JZMLHJRKSJXARY-UHFFFAOYSA-N

Biochem/physiol Actions

Potent and selective inhibitor of Na+/Ca2+ exchange (NCX) acting preferentially at NCX3
YM-244769 hydrochloride is a potent and selective inhibitor of Na+/Ca2+ exchange (NCX) acting preferentially at NCX3. It inhibits the Ca2+ entry mode of NCX more potently than the Ca2+ exit mode. YM-244769 exhibits protection in various hypoxic/ischemic cell injury models with low cell toxicity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Blockade of the forward Na+ /Ca2+ exchanger suppresses the growth of glioblastoma cells through Ca2+ -mediated cell death
Hu HJ, Wang SS, Wang YX, Liu Y, Feng XM, Shen Y, Zhu L, Chen HZ, Song M
British Journal of Pharmacology, 176, 2691-2707 (2019)
YM-244769, a novel Na+/Ca2+ exchange inhibitor that preferentially inhibits NCX3, efficiently protects against hypoxia/reoxygenation-induced SH-SY5Y neuronal cell damage
Iwamoto T, Kita S
Molecular Pharmacology, 70, 2075-2083 (2006)
Kanna Yamashita et al.
Naunyn-Schmiedeberg's archives of pharmacology, 389(11), 1205-1214 (2016-08-03)
Recently, YM-244769 (N-(3-aminobenzyl)-6-{4-[(3-fluorobenzyl)oxy]phenoxy} nicotinamide) has been reported as a new potent and selective Na+/Ca2+ exchange (NCX) inhibitor by using various cells transfected with NCX using the 45Ca2+ fluorescent technique. However, the electrophysiological study of YM-244769 on NCX had not been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service