Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3330

Sigma-Aldrich

MRK-560

≥98% (HPLC)

Synonym(s):

MRK 560, MRK560, N-[cis-4-[(4-Chlorophenyl)sulfonyl]-4-(2,5-difluorophenyl)cyclohexyl]-1,1,1-trifluoromethanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClF5NO4S2
CAS Number:
Molecular Weight:
517.92
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

FC(F)(F)[S](=O)(=O)N[C@@H]1CC[C@@](CC1)([S](=O)(=O)c3ccc(cc3)Cl)c2c(ccc(c2)F)F

InChI

1S/C19H17ClF5NO4S2/c20-12-1-4-15(5-2-12)31(27,28)18(16-11-13(21)3-6-17(16)22)9-7-14(8-10-18)26-32(29,30)19(23,24)25/h1-6,11,14,26H,7-10H2/t14-,18+

InChI key

WDZVWDXOIGQJIO-UJKQEGAGSA-N

Biochem/physiol Actions

MRK-560 is a brain-penetrant, orally active, highly potent and selective gamma-secretase inhibitor (GSI; IC50 = 0.65 nM by SH-SY5Y cellular γ-secretase substrate A4CTF breakdown assay) with little inhibitory potency against a panel of 171 enzymes/receptors. MRK-560 blocks Aβ40 & Aβ42 production with similar potency in SH-SY5Y cultures (IC50 <0.5 nM) and lowers CNS Aβ40 in rats (brain & SCF ED50 in 8h = 6 & 10 mg/kg po.) and APP-YAC mice (brain ED50 in 4h <3 mg/kg po.) in vivo

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service