Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3323

Sigma-Aldrich

MS9715

≥98% (HPLC)

Synonym(s):

(2S,4R)-1-((S)-2-(9-(4-((4-(5-(7-fluoroquinolin-4-yl)-1-methyl-1H-imidazol-4-yl)-3,5-dimethylbenzyl)amino)butanamido)nonanamido)-3,3-dimethylbutanoyl)-4-hydroxy-N-((S)-1-(4-(4-methylthiazol-5-yl)phenyl)ethyl)pyrrolidine-2-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C58H74FN9O5S
Molecular Weight:
1028.33
UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

CC1=C(C(C)=CC(CNCCCC(NCCCCCCCCC(N[C@H](C(N2[C@@H](C[C@H](C2)O)C(N[C@H](C3=CC=C(C=C3)C4=C(N=CS4)C)C)=O)=O)C(C)(C)C)=O)=O)=C1)C5=C(N(C=N5)C)C6=CC=NC7=C6C=CC(F)=C7

InChI

1S/C58H74FN9O5S/c1-36-28-40(29-37(2)51(36)52-53(67(8)34-63-52)46-24-27-61-47-30-43(59)22-23-45(46)47)32-60-25-15-17-49(70)62-26-14-12-10-9-11-13-16-50(71)66-55(58(5,6)7)57(73)68-33-44(69)31-48(68)56(72)65-38(3)41-18-20-42(21-19-41)54-39(4)64-35-74-54/h18-

InChI key

RWXCSOWWDBANDF-LPFSXYLXSA-N

Biochem/physiol Actions

MS9715 is a potent and highly selective degrader (PROTAC) of NSD3 (Nuclear receptor binding SET domain protein 3) that effectively depletes NSD3 and the associated cMyc oncogenic node. MS9715 comprised of NSD3 antagonist BI-9321 linked to an E3 ligase VHL ligand. It potently inhibits growth of NSD3-dependent hematological cancer cells.
Potent and highly selective degrader (PROTAC) of NSD3 that effectively depletes NSD3 and the associated cMyc oncogenic node

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service