Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3322

Sigma-Aldrich

SGC-STK17B-1N

≥98% (HPLC)

Synonym(s):

2-((6-(Benzo[b]thiophen-2-yl)thieno[2,3-d]pyrimidin-4-yl)thio)acetic acid, 2-(6-(Benzo[b]thiophen-2-yl)thieno[2,3-d]pyrimidin-4-ylthio)acetic acid

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$93.80
25 MG
$378.00

$93.80


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$93.80
25 MG
$378.00

About This Item

Empirical Formula (Hill Notation):
C16H10N2O2S3
CAS Number:
Molecular Weight:
358.46
UNSPSC Code:
12352200
NACRES:
NA.77

$93.80


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to very dark grey

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CSC1=C2C=C(C3=CC4=CC=CC=C4S3)SC2=NC=N1)=O

InChI

1S/C16H10N2O2S3/c19-14(20)7-21-15-10-6-13(23-16(10)18-8-17-15)12-5-9-3-1-2-4-11(9)22-12/h1-6,8H,7H2,(H,19,20)

InChI key

RRIRDIKTUQVZCW-UHFFFAOYSA-N

Biochem/physiol Actions

Negative control for STK17B/DRAK2 inhibitor SGC-STK17B-1
SGC-STK17B-1N is a negative control for SGC-STK17B-1 a cell penetrant, potent and selective inhibitor of STK17B/DRAK2 kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service