Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3309

Sigma-Aldrich

DN01

≥98% (HPLC)

Synonym(s):

4-(4-((Dimethylamino)methyl)-2,6-dimethoxyphenyl)-7-fluoro-2-methylisoquinolin-1(2H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23FN2O3
CAS Number:
Molecular Weight:
370.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, White to light brown

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

DN01 is a potent and selective BRD8 first bromodomain inhibitor (BRD8(1) IC50 = 12 nM; BRD4(1) & BRD9 IC50 ≥10 μM) that serves as a useful chemical probe for the selective engagement of BRD8 and enrichment of native NuA4 complexes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemoproteomics Enabled Discovery of Selective Probes for NuA4 Factor BRD8
ACS Chemical Biology, 16(11):, 2185-2192 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service