Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3245

Sigma-Aldrich

dCBP-1

≥98% (HPLC)

Synonym(s):

3-[7-(difluoromethyl)-3,4-dihydro-6-(1-methyl-1H-pyrazol-4-yl)-1(2H)-quinolinyl]-1-[1-[15-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1H-isoindol-5-yl]amino]-1-oxo-4,7,10,13-tetraoxapentadec-1-yl]-4-piperidinyl]-1,4,6,7-tetrahydro-N-methyl-5H-pyrazolo[4,3-c]pyridine-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C51H63F2N11O10
CAS Number:
Molecular Weight:
1028.11
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1CCC(CC1)N2N=C(C3=C2CCN(C3)C(NC)=O)N4C5=C(CCC4)C=C(C6=CN(N=C6)C)C(C(F)F)=C5)CCOCCOCCOCCOCCNC7=CC=C(C8=C7)C(N(C8=O)C9C(NC(CC9)=O)=O)=O

Biochem/physiol Actions

dCBP-1 is a highly potent and selective bifunctional PROTAC degrader of CREB-binding protein (CBP) and p300 (p300/CBP) that potently inhibits enhancer-mediated transcription. dCBP-1 induces complete loss of p300/CBP in multiple cellular models of multiple myeloma.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service