Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SML3199

Sigma-Aldrich

Monomethyl Auristatin E

≥98%

Synonym(s):

MMAE, MMAE toxin, Monomethylauristatin E, N-Methyl-L-valyl-N-[(1S,2R)-4-[(2S)-2-[(1R,2R)-3-[[(1R,2S)-2-hydroxy-1-methyl-2-phenylethyl]amino]-1-methoxy-2-methyl-3-oxopropyl]-1-pyrrolidinyl]-2-methoxy-1-[(1S)-1-methylpropyl]-4-oxobutyl]-N-methyl-L-valinamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H67N5O7
CAS Number:
Molecular Weight:
717.98
UNSPSC Code:
12352200
Product SML3199 is not currently sold in your country. Contact Technical Service

Quality Level

Assay

≥98%

form

film or powder

color

white to off-white

storage temp.

−20°C

SMILES string

N1([C@@H](CCC1)[C@H](OC)[C@@H](C)C(=O)N[C@@H]([C@@H](O)c2ccccc2)C)C(=O)C[C@@H](OC)[C@@H](N(C)C(=O)[C@@H](NC(=O)[C@@H](NC)C(C)C)C(C)C)[C@H](CC)C

InChI

1S/C39H67N5O7/c1-13-25(6)34(43(10)39(49)33(24(4)5)42-38(48)32(40-9)23(2)3)30(50-11)22-31(45)44-21-17-20-29(44)36(51-12)26(7)37(47)41-27(8)35(46)28-18-15-14-16-19-28/h14-16,18-19,23-27,29-30,32-36,40,46H,13,17,20-22H2,1-12H3,(H,41,47)(H,42,48)/t25-,26+,27+,29-,30+,32-,33-,34-,35+,36+/m0/s1

InChI key

DASWEROEPLKSEI-UIJRFTGLSA-N

Biochem/physiol Actions

Monomethyl Auristatin E (MMAE) is a highly potent peptidyl antimitotic agent that blocks the polymerization of tubulin. Monomethyl Auristatin E is a component of a clinically approved antibodydirected conjugates Brentuximab vedotin, Glembatumumab vedotin, Enfortumab vedotin and Polatuzumab vedotin.
highly potent antimitotic agent; anti-tubulin agent;

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral - Aquatic Chronic 3 - Muta. 1B - Repr. 1B - STOT RE 1 - STOT SE 1

Target Organs

male reproductive organs,Bone marrow,thymus,spleen,Liver

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service