Skip to Content
MilliporeSigma
All Photos(1)

Documents

SML3196

Sigma-Aldrich

SU086

≥98% (HPLC)

Synonym(s):

(2E)-3-(3-Nitrophenyl)-1-(2,4,6-trimethoxyphenyl)-2-propen-1-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO6
CAS Number:
Molecular Weight:
343.33
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

SU086 is a cell-permeable, non-toxic, bioavailable chalcone compound that decreases HSP90 levels and impairs glycolysis. SU086 is shown to inhibit prostate cancer cell growth, migration, and invasion in vitro (10 μM) through non-androgen receptor mediated pathways. SU086 displays favorable PK profile and synergizes with enzalutamide and abiraterone to decrease prostate cancer growth in vitro and in vivo. It is a potent inhibitor of selenoprotein thioredoxin reductases (TrxRs).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service